accessibility menu, dialog, popup

UserName

H3COO(S)NH(S)H3CON(S)HOO

CAS RN 75847-73-3

IUPAC Name: (2S)-1-[(2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]pyrrolidine-2-carboxylic acid
Synonyms: enalapril enalaprila
Molecular Weight (g/mol): 376.45
Molecular Formula: C20H28N2O5
InChi Key: GBXSMTUPTTWBMN-XIRDDKMYSA-N
SMILES: CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1CCC[C@H]1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results