missing translation for 'onlineSavingsMsg'
Learn More

Enalapril Maleate 98.0+%, TCI America™

Catalog No. E10101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10101G 1 g
E10105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10101G Supplier TCI America Supplier No. E10101G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76095-16-4
Molecular Formula C24H32N2O9
Molecular Weight (g/mol) 492.53
MDL Number MFCD00133304,MFCD00865774
InChI Key OYFJQPXVCSSHAI-BMSLDGIRSA-N
Synonym enalapril maleate, renitec, glioten, vasotec, baripril, benalapril, crinoren, innovace, naprilene, renivace
PubChem CID 5388961
ChEBI CHEBI:4785
IUPAC Name (2S)-1-[(2S)-2-{[(2R)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]pyrrolidine-2-carboxylic acid; (2Z)-but-2-enedioic acid
SMILES OC(=O)\C=C/C(O)=O.CCOC(=O)[C@@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1CCC[C@H]1C(O)=O

Specifications

Physical Form Powder/Crystals
Color White
Melting Point 144°C
Quantity 1 g
Formula Weight 492.53
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Enalapril Maleate
RTECSNumber TW3666000
TSCA No