missing translation for 'onlineSavingsMsg'
Learn More

Enalapril maleate salt, ≥98% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111015450
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015450 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015450 Supplier Merck Emd Millipore Supplier No. 1661650MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 76095-16-4
Physical Form Neat
Linear Formula C20H28N2O5 · C4H4O4
Synonym (S)-N-[1-(Ethoxycarbonyl)-3-phenylpropyl]-Ala-Pro maleate salt
Type Analytical Standard
Quantity 50 mg
Formula Weight 492.52
Percent Purity ≥98% (TLC)
Grade Analytical Standard