missing translation for 'onlineSavingsMsg'
Learn More

CDS Empore™ RAD Disks

Formerly 3M™ Empore™ Solid Phase Extraction RAD Disks (SPE): 3290 Sr, 3291 Ra, and 3292 Tc. Provide an efficient alternative to conventional radiochemical sample preparation methods that use wet chemistry or packed columns.

$974.68 - $974.68

Specifications

Type Rad Extraction Disk
Quantity 60/Cs.
Length (Metric) 47 mm
View More Specs

Products 1
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
13110007
View Documents
CDS
98040500666EA
For Use in Tc 99 Detection N/A
Only null left
 
Description

Description

  • Molecular recognition technology-Crown Ether bonded Silica particles (12μm)
  • Capture the selected analyte and then also as the counting medium (liquid scintillation or counting)
  • Reduces turnaround time and lowers operating costs
  • Available in 47mm size
Specifications

Specifications

Rad Extraction Disk
60/Cs.
47 mm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'