missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDS Empore™ Anion and Cation Exchange-SR Extraction Disks
Formerly 3M™ Empore™ Anion and Cation-SR Solid Phase Extraction Disks (SPE): 2252, 2251 & 2352. Provide significantly faster extraction time with no negative effect on recoveries
Supplier: CDS 98060402298EA
Description
- Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
- Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
- Available with a variety of packings in 47 and 90 mm sizes
- Anion exchange disks are approved for use in EPA Methods 549.1 and 552.1
- Replace liquid/solid procedures for the determination of haloacetic acids in water, as well as chlorinated herbicide Dalapon™
Specifications
| EPA Methods 549.1 and 552.1 | |
| Analytical Preparation of Aqueous Environmental Samples | |
| Extraction Disk | |
| 90% | |
| 4.7 cm |
| Anion Exchange | |
| SAX | |
| 60/Cs. | |
| 4.7 cm |