missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Emerin Recombinant Rabbit Monoclonal Antibody (23GB5320)

Catalog No. MA553885
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553885 20 μL
MA553884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553885 Supplier Invitrogen™ Supplier No. MA553885
Only null left

Rabbit Recombinant Monoclonal Antibody

Emerin Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Xeroderma pigmentosum type G (XPG) is a human genetic disease exhibiting extreme sensitivity to sunlight. The XPG protein, a member of the flap endonuclease 1 (FEN-1) structure-specific DNA repair endonuclease family, is an enzyme essential for DNA repair of the major kinds of solar ultraviolet (UV)-induced DNA damages. Human XPG nuclease makes the 3' incision during nucleotide excision repair of DNA. The enzyme cleaves model DNA bubble structures specifically near the junction of unpaired DNA with a duplex region. A 29-amino acid region of human XPG (residues 981-1009) contains the PCNA binding activity. A conserved arginine in XPG (Arg992) is crucial for its PCNA binding activity. Replication Protein A (RPA) binds specifically and directly to two excision repair proteins, the xeroderma pigmentosum damage-recognition protein XPA and the endonuclease XPG.

Specifications

Antigen Emerin
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5320
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Emd
Gene Accession No. O08579, P50402, Q63190
Gene Alias AW550900; EDMD; EMD; Emerin; LEM domain containing 5; LEMD5; STA
Gene Symbols Emd
Host Species Rabbit
Immunogen A synthesized peptide derived from human Emerin (200-254AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13726, 2010, 25437
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less