Learn More
MilliporeSigma™ Washer Distribution Kit
For installation on the left/right side of a pure water storage reservoir
$2,270.00
Specifications
For Use With (Equipment) Pure Water Storage Reservoir
Type Washer Distribution Kit
Voltage 115 V
View More Specs
Products 2
missing translation for 'productCode' missing translation for 'brand' For Use With (Application) Price Quantity & Availability  
missing translation for 'productCode' missing translation for 'brand' For Use With (Application) Price Quantity & Availability  
ZWDK6L1US
View Documents Promotion Details
MilliporeSigma™
ZWDK6L1US
Left Side Installation
Each for $2,270.00
Only null left
ZWDK6R1US
View Documents Promotion Details
MilliporeSigma™
ZWDK6R1US
Right Side Installation
Each for $2,270.00
Only null left
Description
Description
  • Provides pressurized flow (15 to 16.2L/min. at 1 bar (15psi)) of Elix™ or RiOs™ water from a reservoir to a glassware washer or other laboratory appliance on-demand via a pressure switch
  • 115/230V
Specifications
Specifications
Pure Water Storage Reservoir
Washer Distribution Kit
115 V