Learn More
MilliporeSigma™ Washer Distribution Kit
Catalog No. ZWDK6L1US
Change view
Click to view available options
For Use With (Application):
Left Side Installation
Right Side Installation
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
ZWDK6L1US Left Side Installation
ZWDK6R1US Right Side Installation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ZWDK6L1US Supplier MilliporeSigma™ Supplier No. ZWDK6L1US
For installation on the left/right side of a pure water storage reservoir
  • Provides pressurized flow (15 to 16.2L/min. at 1 bar (15psi)) of Elix™ or RiOs™ water from a reservoir to a glassware washer or other laboratory appliance on-demand via a pressure switch
  • 115/230V
Specifications
For Use With (Equipment) Pure Water Storage Reservoir
Type Washer Distribution Kit
For Use With (Application) Left Side Installation
Voltage 115 V