missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RIPAb+™ Upf1 RIP Validated Antibody and Primer Set

Catalog No. 03191MI
Change view
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03191MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03191MI Supplier MilliporeSigma™ Supplier No. 03191

This RIPAb+ Upf1 -RIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.

Includes negative control normal rabbit IgG antibody and control primers specific for the cDNA of human ARF1.

Specifications

Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Gene Accession No. NP_002902.2
Gene Symbols pNORF1, EC 3.6.1., FLJ43809, FLJ46894, HUPF1, KIAA0221, NORF1, RENT1, hUpf1
Host Species Rabbit
Immunogen KLH-conjugated synthetic linear peptide corresponding to Upf1.
Purification Method Affinity Purfied
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Bovine, Canine, Human, Mouse, Rat
Content And Storage -20°C for 12 months.
Product Type RIPAb+ Upf1 RIP Validated Antibody and Primer Set
Form Purified
Show More Show Less