missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RIPAb+™ STAU1 (Staufen 1) RIP Validated Antibody and Primer Set

Catalog No. 03116MI
Change view
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03116MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03116MI Supplier MilliporeSigma™ Supplier No. 03116

This RIPAb+ STAU1 (Staufen 1) -RIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.

Includes negative control normal rabbit IgG antibody and control primers specific for the human ARF1 mRNA.

Specifications

Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Description Negative control normal rabbit IgG antibody and control primers specific for the human ARF1 mRNA
Gene Accession No. NP_001032405
Host Species Rat
Immunogen Synthetic peptide from amino acids 2-51 of human STAU 1.
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Human
Content And Storage -20°C for 12 months.
Product Type RIPAb+™ STAU1 (Staufen 1) RIP Validated Antibodies
Form Purified
Show More Show Less