missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RIPAb+™ SMN RIP Validated Antibody and Primer Set

Catalog No. 03178MI
Change view
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03178MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03178MI Supplier MilliporeSigma™ Supplier No. 03178

This RIPAb+ SMN -RIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.

Includes negative control mouse IgG antibody and control primers specific for the cDNA of human U1snRNP.

Specifications

Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Description Negative control mouse IgG antibody and control primers specific for the cDNA of human U1snRNP
Gene Accession No. NP_000335.1
Gene Symbols SMNT, SMN1, BCD541, Gemin-1, SMA, SMA1, SMA2, SMA3, SMA4, SMA™, SMN, SMNC
Host Species Mouse
Immunogen Histidine-tagged recombinant protein corresponding to human SMN.
Purification Method Protein G purfied
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Human, Mouse, Xenopus species
Content And Storage -20°C for 12 months.
Product Type RIPAb+ SMN RIP Validated Antibody and Primer Set
Form Purified
Isotype IgG1 κ
Show More Show Less