missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RIPAb+™ PUM2 RIP Validated Antibody and Primer Set

Catalog No. 03241MI
Change view
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03241MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03241MI Supplier MilliporeSigma™ Supplier No. 03241

Detect RIPAb+ PUM2 using this RIPAb+ PUM2 Antibody validated for use in ICC, IHC(P), Western Blotting, RNA Binding Protein Immunoprecipitation (RIP).

Detect RIPAb+ PUM2 using this RIPAb+ PUM2 Antibody validated for use in immunocytochemistry, immunohistochemistry (paraffin), Western Blotting, RNA Binding Protein Immunoprecipitation (RIP).

Specifications

Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Gene Accession No. NP_056132
Gene Symbols PUMH2, PUM2, PUML2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the N-terminus of human Pumilio-2.
Purification Method Affinity Purfied
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Canine, Chimpanzee, Human, Macaque, Mouse, Rhesus Monkey
Content And Storage -20°C for 12 months.
Product Type RIPAb+ PUM2 RIP Validated Antibody
Form Purified
Isotype IgG2a
Show More Show Less