missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RIPAb+™ pan Ago RIP Validated Antibody and Primer Set

Catalog No. 03248MI
Change view
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03248MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03248MI Supplier MilliporeSigma™ Supplier No. 03248

This RIPAb+ pan Ago -RIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.

Includes negative control normal mouse IgG antibody and control primers specific for the cDNA of human IGF2 mRNA.

Specifications

Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Gene Accession No. NP_001158095
Gene Symbols KIAA1567, EIF2C2l, AGO2, Q10, PPD, eIF-2C 2, hAgo2, Argonaute1, Ago1, eIF-2C 1, EIF2C1, AGO1, Argonaute3, AGO3, EIF2C3, EIF2C4, : AGO4
Host Species Mouse
Immunogen Histidine - tagged recombinant protein corresponding to human Ago.
Purification Method Protein G purfied
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Human
Content And Storage -20°C for 12 months.
Product Type RIPAb+ pan Ago RIP Validated Antibody and Primer Set
Form Purified
Isotype IgG1 κ
Show More Show Less