missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RIPAb+™ hnRNPA1 RIP Validated Antibody and Primer Set

Catalog No. 03204MI
Change view
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03204MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03204MI Supplier MilliporeSigma™ Supplier No. 03204

This RIPAb+ hnRNPA1 -RIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.

Includes negative control mouse IgG antibody and control primers specific for the mRNA of human E2F3.

Specifications

Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Gene Accession No. NP_002127
Gene Symbols hnRNPA1, HNRNPA1, HNRPA1
Host Species Mouse
Immunogen Full length native protein partially purified human hnRNP A1.
Purification Method Protein G purfied
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Human
Content And Storage -20°C for 12 months.
Product Type RIPAb+ hnRNPA1 RIP Validated Antibody and Primer Set
Form Purified
Isotype IgG2b κ
Show More Show Less