missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ RIPAb+™ Fragile X Mental Retardation RIP Validated Antibody and Primer Set
Description
Includes negative control mouse IgG antibody and control primers specific for mouse Iqsec1.
Spécifications
Spécifications
| Applications | ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Aperçu général | Negative control mouse IgG antibody and control primers specific for mouse Iqsec1 |
| Numéro d’acquisition du gène | NP_002015.1 |
| Symboles de gène | FMRP, FMR1 |
| Espèces hôtes | Mouse |
| Immunogène | Fusion protein with a full length FMRP (human). |
| Quantité | 10 Assays/Set (approx. 5μg of antibody per RIP) |
| Espèces cibles | Human, Mouse, Rat |
| Contenu et stockage | -20°C for 12 months. |
| Afficher plus de résultats |