missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ RIPAb+™ EF1α RIP Validated Antibody and Primer Set
Description
Includes negative control mouse IgG antibody.
Spécifications
Spécifications
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Description | Negative control mouse IgG antibody and control primers |
| Gene Accession No. | NP_001393.1 |
| Gene Symbols | LENG7, EEF1A1, EEF1A, EF1A |
| Host Species | Mouse |
| Immunogen | Crude calmodulin-binding proteins from Trypanosoma brucei isolated using calmodulin-affinity chromatography |
| Quantity | 10 Assays/Set (approx. 5μg of antibody per RIP) |
| Target Species | Human, Mammalia, Yeast (S. cerevisiae) |
| Content And Storage | -20°C for 12 months. |
| Afficher plus de résultats |