missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RIPAb+™ EED RIP Validated Antibody and Primer Set

Numéro de catalogue. 03196MI
Modifier l'affichage
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
03196MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 03196MI Fournisseur MilliporeSigma™ Code fournisseur 03196

This RIPAb+ EED -RIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.

Includes negative control normal mouse IgG antibody and control primers specific for the cDNA of human lincRNA SFPQ.

Spécifications

Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Gene Accession No. NP_068676
Gene Symbols hEED, HEED, WAIT-1, WAIT1
Host Species Mouse
Immunogen Recombinant fusion protein of full length murine EED tagged with MBP.
Purification Method Protein G purfied
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Human, Mouse
Content And Storage -20°C for 12 months.
Product Type RIPAb+ EED RIP Validated Antibody and Primer Set
Form Ascites
Isotype IgG2a κ
Afficher plus de résultats Afficher moins de résultats