missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ RIPAb+™ AUF1 RIP Validated Antibody and Primer Set
Description
Includes negative control rabbit IgG antibody and control primers specific for human FOS.
Spécifications
Spécifications
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Description | Negative control rabbit IgG antibody and control primers specific for human FOS |
| Gene Accession No. | NP_001003810.1 |
| Gene Symbols | hnRNP D0, HNRNPD, AUF1, HNRPD |
| Host Species | Rabbit |
| Immunogen | Human AUF1 purified by Ni2+ affinity column |
| Quantity | 10 Assays/Set (approx. 5μg of antibody per RIP) |
| Target Species | Human, Mouse, Rat |
| Content And Storage | -20°C for 12 months. |
| Afficher plus de résultats |