missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ RIPAb+™ Ago2 RIP Validated Antibody and Primer Set
Description
Includes negative control mouse IgG antibody and control primers specific for human FOS.
Spécifications
Spécifications
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Description | Negative control mouse IgG antibody and control primers specific for human FOS |
| Gene Accession No. | NP_001158095.1 |
| Gene Symbols | PPD, Ago2, eIF-2C 2, eIF2C 2 |
| Host Species | Mouse |
| Immunogen | KLH-conjugated, synthetic peptide containing the sequence YSGAGPALAPPAPPPPIQG |
| Quantity | 10 Assays/Set (approx. 5μg of antibody per RIP) |
| Target Species | Bovine, Human |
| Content And Storage | -20°C for 12 months. |
| Afficher plus de résultats |