missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Upstate™ Recombinant Active PCAF Protein
Specifications
Specifications
| For Use With (Application) | Histone Acetyltransferase Assays |
| Gene ID (Entrez) | NM_003884.3 |
| Name | Acive PCAF Protein |
| Quantity | 50 μg |
| Source | E. coli |
| Storage Requirements | -20°C for 6 months |
| Recombinant | Recombinant |