Learn More
MilliporeSigma™ Upstate™ Rac/cdc42 Assay Reagent (PAK1 PBD, Agarose)
Supplier: MilliporeSigma™ 14325
Specifications
| Affinity Binding Assay | |
| NM_044472.2, NM_001039802.1, NM_001791.3 | |
| 300 μg | |
| -20°C for 12 months. | |
| Human |
| Bound to glutathione agarose | |
| PAK1 PBD fusion GST-tagged fusion protein | |
| E. coli | |
| GST-Tag | |
| Native |