missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate Endothelial Cell Growth Supplements

Catalog No. 02101MI
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02101MI 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02101MI Supplier MilliporeSigma™ Supplier No. 02101

Designed for use as a cell culture media supplement

ECGS is mitogenic for many types of cells under reduced or serum-free conditions, such as mammalian, avian and human endothelial cells, smooth muscle cells, keratinocytes, melanocytes and hybridomas; ECGS often fully substitutes for feeder layers in culture of fastidious cells.

Specifications

Content And Storage 2 years at 4°C; rehydrated: 3 months at -20°C
For Use With (Application) Cell Culture
Quantity 50 mg