Learn More
MilliporeSigma™ Upstate ChIPAb+ REST - ChIP Validated Antibody and Primer Set
REST purified antibody against GST fusion protein corresponding to amino acids 801-1097 of human REST
Supplier:  MilliporeSigma™ 17641
Catalog No. 17-641-MI
Only null left
Includes: ChIP-grade antibody and the specific control PCR primers used for chromatin immunoprecipitation of REST
Description
Description
Specifications
Specifications
XBR, NRSF
ChIPAb+ REST - ChIP Validated Antibody and Primer Set
Product Suggestions
Product Suggestions