missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Upstate Bovine Pituitary Extract
Specifications
Specifications
| Content And Storage | Stable for 2 years at 4°C from date of shipment. Rehydrated: Stable for 3 months at -20°C |
| For Use With (Application) | Cell Culture |
| Quantity | 150 mg |