missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Tubing

Catalog No. XT1200000
Change view
Click to view available options
Overall Length (Metric):
70 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Overall Length (Metric)
XT1200000 70 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. XT1200000 Supplier MilliporeSigma™ Supplier No. XT1200000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PTFE Tubing for solvent/buffer

Specifications

For Use With MilliSolve Filtration System
Type Tubing
Color Clear
Diameter (English) Outer 1.4 in.
Diameter (Metric) Outer 3.1 cm
Overall Length (English) 27.5 in.
Overall Length (Metric) 70 cm
Material PTFE