missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate™ Trimethyl-Histone H3 (Lys36)α, Rabbit Monoclonal, ChIP Validated Antibody and Primer Set

Numéro de catalogue. 1710032MI
Modifier l'affichage
Click to view available options
Quantité:
25 Assays
Conjugué:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Conjugué
1710032MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 1710032MI Fournisseur MilliporeSigma™ Code fournisseur 1710032

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Monoclonal Antibody

Specifically detects Trimethyl-Histone H3 (Lys36)α Clone: in Chicken, Human samples, and it is validated for ChIP, Dot Blot, Functional Assay, Western Blotting

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome, the basic subunit of chromatin. Histones are modified post-translationally by the actions of enzymes in both the nucleus and cytoplasm. These modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. These modifications can alter local chromatin architecture, or recruit trans-acting factors that recognize specific histone modifications (the histone code hypothesis). The modifications occur predominantly on the N-terminal and C-terminal tails that extend beyond the nucleosome core particle. Methylation of histone H3 on Lys36 (H3K36me2/3) is tightly associated with actively transcribed genes, and this modification is found primarily within the coding region, suggesting H3K36 methylation is necessary for efficient RNA polymerase II elongation and processivity.

Spécifications

Antigène Trimethyl-Histone H3 (Lys36)α
Applications ChIP Assay, Dot Blot, Functional Assay, Western Blot
Classification Monoclonal
Conjugué Unconjugated
Formule Anti-Trimethyl-Histone H3 (Lys36) (rabbit monoclonal IgG). One vial containing 50μL of protein A purified IgG in a solution containing 0.07 M Tris-glycine, 0.105 M NaCl, pH 7.4, 0.035% sodium azide and 30% glycerol. Normal Rabbit IgG. One vial containing 125μg Rabbit IgG in 125μL of storage buffer containing 0.05% sodium azide. ChIP Primers, BDNF Intron. One vial containing 75μL of 5μM of each primer specific for human BDNF intron. FOR: ACCCCAACCTCTAACAGCATTA; REV: TGTCTCTCAGCAGTCTTGCATT
Numéro d’acquisition du gène Q16695
Symboles de gène H3.4, H3/g, H3/t, H3FT, H3T, H3t, MGC126886, MGC126888, OTTHUMP00000037945
Espèces hôtes Rabbit
Immunogène KLH-conjugated, synthetic peptide containing the sequence ....GVme3KKP…, in which me3K corresponds to human trimethyl-histone H3 (Lys36).
Méthode de purification Protein A purified
Quantité 25 Assays
Statut réglementaire RUO
Disciplines de recherche Epigenetics, Nuclear Function
Primaire ou secondaire Primary
ID du gène (Entrez) NP_003484
Spécifications de test Recognizes Trimethyl-Histone H3 (Lys36), Mr ∽17kDa.
Comprend This ChIPAb+ Trimethyl-Histone H3 (Lys36) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Espèces cibles Chicken, Human
Contenu et stockage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Forme Purified
Isotype IgG
Afficher plus de résultats Afficher moins de résultats

For Research Use Only