missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Transferrin, Holo, Human Plasma, Protein
Description
Native holotransferrin from human plasma. Transferrin is a serum protein involved in iron binding and transport.
Specifications
Specifications
| Formulation | Lyophilized from 150mM NaCl, 20mM sodium phosphate buffer, pH 7.4. |
| Preparation Method | H2O (10mg/mL) |
| Purification Method | SDS-PAGE |
| Quantity | 500 mg |
| Immunogen | Transferrin, Holo |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C) for long-term storage or add NaN3 and refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 2 weeks at 4°C. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Lyophilized |
| Show More |