missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Steripak™-GP Sterile Filter Units

Catalog No. p-105804
Change view
Click to view available options
Volume (Metric):
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
SPGPM10RJ 10 mL
SPGPM20RJ 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SPGPM10RJ Supplier MilliporeSigma™ Supplier No. SPGPM10RJ
Only null left

Includes

6mm (0.25 in. ) stepped hose barb inlet, 6mm (0.25 in. ) stepped hose barb outlet and filling bell.

Fast flow and high throughput

Steripak-GP filter units are designed for larger scale pressure-driven filtration of tissue culture media with or without serum. They are single use and are not autoclavable. The filter units come in two sizes, with the smaller unit able to filter up to 10 L and the larger up to 20 L. Both sizes utilize the low protein binding, fast flow 0.22 μm Millipore Express membrane. Each unit has an integral filling bell for bottle-top use and a manually operated upstream air vent to prevent membrane air locking. They are supplied sterile and ready to connect to a pump or pressure vessel (not included).

Steripak™-GP filter units are designed for larger scale, pressure-driven filtration of tissue culture media with or without serum. Each filter is for a single use and is not autoclavable. The filter units come in two sizes with the smaller unit able to filter up to 10 L and the larger up to 20 L.

Features & Benefits:
  • Millipore Express™ PLUS membrane for fastest flow rates and low protein binding
  • Larger scale, pressure-driven filtration device for faster flow rates speeds time-consuming, large-volume filtrations
  • 10 L and 20 L capacity sizes for maximum laboratory-scale filtration options
  • Manually operated top air vent to prevent air lock stop of flow
  • Filling bell for cleaner filling environment around the opening of the receptacle bottle

Applications:
Sterilization of Aqueous Solutions, Media Preparation, Buffer Preparation

Specifications

Disposable Single-use
Material SAN
Material (Membrane) PES
Pore Size 0.22 μm
Sterility Sterile
Max. Inlet Pressure 75 psi forward, 5 psi reverse
No. per Case 3Pack
Outlet Fitting 6 mm stepped hose barb, filling bell
Outlet Style 6mm stepped hose barb, filling bell
Temperature (English) Operating 113°F
Temperature (Metric) Operating 45°C
Type Bottle Top Filter
Volume (Metric) 10 mL
Height (English) 2.99 in.
Height (Metric) 7.6 cm
Diameter (Metric) Filter 9.9 cm
Show More Show Less