Learn More
MilliporeSigma™ Accessories and Replacement Parts In-Line Stainless Steel Filter Holders 47 mm
Catalog No. p-322751
Change view
Click to view available options
Type:
Back-pressure Screen
Filter Support Screen
O-Ring
Material:
Silicone
Stainless Steel
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Material
XX4404703 Back-pressure Screen Stainless Steel
XX4304701 O-Ring Silicone
XX4504704 Filter Support Screen Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. XX4404703 Supplier MilliporeSigma™ Supplier No. XX4404703
Only null left
Accessories for filter holders for sample preparation.
XX4204709, XX4304701, XX4404703, 5614, and 00907P are replacement parts for Filter Holder XX4404700
SS Filter, Depends on filter type used.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) In-Line Stainless Steel Filter Holder (47 mm)
Material Stainless Steel
Sterility Non-sterile
Type Back-pressure Screen
Diameter (Metric) Inside 47 mm