missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ SOST, Mouse, Recombinant Protein

Catalog No. 56769325UG
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56769325UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56769325UG Supplier MilliporeSigma™ Supplier No. 56769325UG
Recombinant, mouse sclerostin (SOST) consisting of amino acids 24-211, expressed in NS0-derived murine myeloma cells with an N-terminal His·Tag™. SOST is expressed by osteoclasts in developing bones of mouse embryos and acts as a negative regulator of bone formation through repression of BMP-induced osteogenesis.

Specifications

Formulation Lyophilized from 0.2μM filter-sterilized PBS, BSA.
Preparation Method Reconstitute to 200μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Purification Method SDS-PAGE
Quantity 25 μg
Immunogen SOST
Storage Requirements Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C.
Regulatory Status RUO
Species Mouse
Recombinant Recombinant
Form Solid
Purity or Quality Grade ≥90%
Show More Show Less