missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Sodium Butyrate Solution

Catalog No. TR1008G
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TR1008G 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TR1008G Supplier MilliporeSigma™ Supplier No. TR1008G

enhances the transient expression of plasmids containing the SV40 early promoter in transfected cells.

This reagent enhances the transient expression of plasmids containing the SV40 early promoter in transfected cells. Available as a 500mM stock solution (1mL). A complete protocol is included with shipment.

Specifications

Content And Storage Store at 2-8° C for 1 year
For Use With (Application) Enhances transient expression of plasmids containing SV40 early promoter in transfected cells
Quantity 1 mL