missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ SNAP i.d. 2.0™ Protein Detection Systems
For Immunohistochemistry (Tissue), Immunohistochemistry, Immunohistochemistry (Paraffin)
$3764.10 - $5030.77
Specifications
| Detection Method | Chemiluminescence, Chromogenic, Fluorescence |
|---|---|
| For Use With (Application) | Immunohistochemistry |
| Includes | Base, IHC frame and incubation cover, Assembly fixture, Vacuum tubing and Quick start guide |
| Quantity | 1/Pk. |
| Storage Conditions | Room Temperature |
Includes: Base, IHC frame and incubation cover, Assembly fixture, Vacuum tubing and Quick start guide
Description
- Unique vacuum technology evenly removes solutions and no more dipping, pouring
- Detection Method: Chemiluminescence Chromogenic Fluorescence
- Frame and slide holders enable to process up to 12 tissue slides
Specifications
| Chemiluminescence, Chromogenic, Fluorescence | |
| Base, IHC frame and incubation cover, Assembly fixture, Vacuum tubing and Quick start guide | |
| Room Temperature |
| Immunohistochemistry | |
| 1/Pk. |