Learn More
MilliporeSigma™ SNAP i.d. 2.0™ IHC Frame
For use with SNAP i.d. 2.0 protein detection system
Supplier:  MilliporeSigma™ SNAP2FRIHC
Numéro de catalogue. SNAP2FRIHC
Il en reste null
Découvrez plus d'offres spéciales
Comprend: Incubation cover and assembly fixture
Description
Description
  • Enables to process up to 12 tissue slides
  • For use with immunohistochemistry slides
Spécifications
Spécifications
Incubation cover and assembly fixture
1/Pk.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS