Learn More
MilliporeSigma™ RoClean™ Cleaning Pouches for Water Purification Systems
Keep your purification system clean with easy-to-use RoClean pouches.
$315.91
Specifications
| For Use With (Equipment) | Elix™, Milli-RO™, Milli-RX™ and Millipore™ RiOs™ water purification system |
|---|
Description
- For cleaning RO membrane in Elix™, Milli-RO™, Milli-RX™, and low-volume Millipore™ RiOs™ Water Purification Systems
- Available in acidic (ROClean A) and basic (ROClean B) formulas
†For details, see Catalog 2004/05.
Specifications
| Elix™, Milli-RO™, Milli-RX™ and Millipore™ RiOs™ water purification system |