missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RIPAb+™ IGF2BP2 RIP Validated Antibody and Primer Set

Catalog No. 03251MI
Change view
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03251MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03251MI Supplier MilliporeSigma™ Supplier No. 03251

This RIPAb+ IGF2BP2 -RIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.

IGF2BP2 (Insulin-like growth factor 2 mRNA-binding protein 2) is a member of the IGF-II mRNA-binding protein family that post-transcriptionally regulates IGF2 during developmental stages.

Specifications

Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Concentration 0.176mg/mL
Gene Accession No. NP_001007226
Gene Symbols VICKZ2, IGF2BP2, IMP2
Host Species Mouse
Immunogen Histidine-tagged recombinant protein corresponding to human IGF2BP2
Purification Method Protein G purfied
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Human
Content And Storage -20°C for 12 months.
Product Type RIPAb+ IGF2BP2 Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less