missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RIPAb+™ Ago1 RIP Validated Antibody and Primer Set

Catalog No. 03249MI
Change view
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03249MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03249MI Supplier MilliporeSigma™ Supplier No. 03249

This RIPAb+ Ago1 -RIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.

Includes normal mouse IgG and primers specific for human IGF2 mRNA.

Specifications

Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Gene Accession No. NP_036331
Gene Symbols AGO1, EIF2C1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Ago1
Purification Method Protein G purfied
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Human
Content And Storage -20°C for 12 months.
Product Type RIPAb+ Ago1 Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less