missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PKG, Iα, Bovine Lung, Protein

Catalog No. 5395784UG
Change view
Click to view available options
Quantity:
4 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5395784UG 4 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5395784UG Supplier MilliporeSigma™ Supplier No. 5395784UG
Only null left
Native PKGIα purified from bovine lung. PKG is involved in regulation of smooth muscle relaxation, platelet function, and cell division. PKG is activated by binding of cGMP to regulatory subunits within the holoenzyme, but in contrast to PKA, the activation of PKG does not invlove physical dissociation of catalytic from the regulatory domains. The activated enzyme phosphorylates target proteins at serine and/or threonine residues that are found within the recognition motif, Arg-Lys-Arg-Ser-Arg-Ala-Glu.

Specifications

Concentration Concentration on Label
Formulation In 150mM NaCl, 25mM β-mercaptoethanol, 10mM K2HPO4, 1mM EDTA, 12% sucrose, pH 6.8.
Quantity 4 μg
Immunogen PKG, Iα
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Bovine
Recombinant Native
Form Liquid