missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ pIEx-7 Ek/LIC Vector Kit

Catalog No. 713393
Change view
Click to view available options
Quantity:
20 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
713393 20 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 713393 Supplier MilliporeSigma™ Supplier No. 713393

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The pIEx™-7 Ek/LIC Vector is a ligation-independent cloning-prepared derivative of the pIEx-6 vector designed to allow rapid, high-level protein expression in insect cells without the time-consuming process of creating a recombinant baculovirus. This vector features the hr5 enhancer and the IE1 (immediate early) promoter to direct expression in insect cells and a minimal N-terminal His·Tag™ coding sequence. This promoter/enhancer combination recruits endogenous insect cell transcription machinery, thereby avoiding baculovirus infection and the associated cytotoxic effects.

Specifications

Promoter IE1
Content And Storage Store at or below -70°C.
Quantity 20 Reactions
For Use With (Application) Rapid, high-level protein expression in insect cells without creating a recombinant baculovirus

For Research Use Only