missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ pIEx-2 DNA Vector

Catalog No. 712383
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
712383 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 712383 Supplier MilliporeSigma™ Supplier No. 712383

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The pIEx™ vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells. Transcription is driven by the AcNPV-derived hr5 enhancer and immediate early promoter, IE1. pIEx-2 contains N-terminal GST·Tag, His·Tag, and S·Tag coding sequences, and a C-terminal HSV·Tag coding sequence.

Specifications

Content And Storage Store at -20°C.
Quantity 20 μg
For Use With (Application) Cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells

For Research Use Only