missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Phosphatase, Acid, Potato, Protein
Description
Native potato acid phosphatase. May be used for the removal of phosphate groups from phosphoproteins, such as β-casein, ovalbumin, and pepsinogen.
Specifications
Specifications
| Preparation Method | 3.2M ammonium sulfate, pH 6.0, containing 1% BSA or 6% BSA in H2O |
| Quantity | 1 kU |
| Immunogen | Phosphatase, Acid |
| Storage Requirements | Following reconstitution, refrigerate (4°C). Stock solutions in ammonium sulfate are stable for up to 4 months at 4°C. Stock solutions in aqueous BSA should be used immediately upon reconstitution. |
| Regulatory Status | RUO |
| Species | Potato |
| Recombinant | Native |
| Form | Lyophilized |