missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ pET Expression System 22b

Catalog No. 707653
Change view
Click to view available options
Quantity:
Sufficient for up to 10 transformations in each host
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
707653 Sufficient for up to 10 transformations in each host
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 707653 Supplier MilliporeSigma™ Supplier No. 707653

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The pET-22b vector carries an N-terminal pelB signal sequence for potential periplasmic localization, plus optional C-terminal His·Tag™ sequence. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map, TB038VM.

Specifications

Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity Sufficient for up to 10 transformations in each host

For Research Use Only