missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ pET-50b(+) DNA Vector

Catalog No. 714643
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
714643 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 714643 Supplier MilliporeSigma™ Supplier No. 714643
The pET-50b(+) vector is designed for cloning and high-level expression of target proteins fused with the 495 aa NusA (Nus·Tag™) protein that is cleavable with the human rhinovirus (HRV) 3C protease.

Specifications

Content And Storage Store at or below -70°C.
Quantity 10 μg
For Use With (Application) pET-50b(+) Vector (10 μg); Induction control strain (supplied as a glycerol stock)

For Research Use Only