missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ pET-44b(+) DNA Vector

Catalog No. 711233
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
711233 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 711233 Supplier MilliporeSigma™ Supplier No. MIL711233

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The pET-44 vectors are designed for cloning and high-level expression of polypeptide sequences fused with the 495 aa NusA (Nus·Tag™) protein. The solubility of many target proteins in the E. coli cytoplasm is increased when they are expressed NusA fusions.

Specifications

Content And Storage Store at or below -70°C.
Quantity 10 μg
For Use With (Application) Cloning and high-level expression of polypeptide sequences fused with the 495 aa NusA (NusTag™) protein

For Research Use Only