Learn More
MilliporeSigma™ pET-29b(+) DNA Vector
Catalog No. 698723
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
698723 10 μg
1 options
Catalog No. 698723 Supplier MilliporeSigma™ Supplier No. 698723
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The pET-29a-c(+) vectors carry an N-terminal S·Tag™/thrombin configuration plus an optional C-terminal His·Tag™ sequence. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map.
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 10 μg
For Research Use Only