missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Pentraxin 3/TSG-14, Mouse, Recombinant Protein
Description
Recombinant, mouse pentraxin 3, consisting of amino acids 18-381 and expressed in NS0-derived murine myeloma cells with a C-terminal His·Tag™. Pentraxin 3, a secreted glycoprotein, exhibits a C-terminal Pentraxin domain and a unique N-terminal domain. Its increased expression may enhance the immune response to protect a host from infection.
Specifications
Specifications
| Formulation | Lyophilized from 0.2μM filtered MOPS, NaCl, CaCl2, BSA. |
| Preparation Method | Reconstitute to 100μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin. |
| Purification Method | SDS-PAGE |
| Quantity | 25 μg |
| Immunogen | Pentraxin 3/TSG-14 |
| Storage Requirements | Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C. |
| Regulatory Status | RUO |
| Species | Mouse |
| Recombinant | Recombinant |
| Form | Solid |
| Show More |