Learn More
MilliporeSigma™ PEI Cellulose F Coated TLC Sheets
For analysis of polar substances with exchange-active groups
Supplier:  MilliporeSigma™ 1.05579.0001
 View more versions of this product
Catalog No. M1055790001
Only null left
Description
Description
  • Standard separations based on microcrystalline cellulose
  • Polyethylenimine-modified, and acts as strong basic anion exchanger
  • Provided with fluorescent indicator with exitation wavelength 254/366nm for UV detection
  • Fibre case packaging
  • 2-dimensional separations such as amino acid “fingerprints”
  • Metabolic studies
Specifications
Chemical Identifiers
60 Å
Cellulose TLC Plate
Analysis of Polar Substances, Separation of Hydrophilic Substances by Partition Chromatography, for Investigating in-Vitro Samples Derived From The Human Body Without Any Medical Objective
20 x 20 cm
20 cm
25/Pk.
254 and 366 nm
100 μm
20 cm
Specifications
Plastic
25/Pk.
254 and 366 nm
PEI Cellulose F
20 x 20 cm
20 cm
60 Å
Cellulose TLC Plate
Analysis of Polar Substances, Separation of Hydrophilic Substances by Partition Chromatography, for Investigating in-Vitro Samples Derived From The Human Body Without Any Medical Objective
100 μm
20 cm
Product Suggestions
Product Suggestions