Learn More
MilliporeSigma™ PEI Cellulose F Coated TLC Sheets
For analysis of polar substances with exchange-active groups
Supplier: MilliporeSigma™ 1.05574.0001
Description
- Standard separations based on microcrystalline cellulose
- Polyethylenimine-modified, and acts as strong basic anion exchanger
- Provided with fluorescent indicator with exitation wavelength 254/366nm for UV detection
- Fibre case packaging
- 2-dimensional separations such as amino acid “fingerprints”
- Metabolic studies
Chemical Identifiers
| 60 Å | |
| Cellulose TLC Plate | |
| Analysis of Polar Substances, Separation of Hydrophilic Substances by Partition Chromatography, for Investigating in-Vitro Samples Derived From The Human Body Without Any Medical Objective | |
| 20 x 20 cm | |
| 20 cm |
| 25/Pk. | |
| 254 and 366 nm | |
| 100 μm | |
| 20 cm |
Specifications
| Aluminum | |
| 25/Pk. | |
| 254 and 366 nm | |
| Cellulose F | |
| 20 x 20 cm | |
| 20 cm |
| 60 Å | |
| Cellulose TLC Plate | |
| Analysis of Polar Substances, Separation of Hydrophilic Substances by Partition Chromatography, for Investigating in-Vitro Samples Derived From The Human Body Without Any Medical Objective | |
| 100 μm | |
| 20 cm |