missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ pACYCDuet-1 DNA Vector

Catalog No. 711473
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
711473 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 711473 Supplier MilliporeSigma™ Supplier No. MIL711473

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

pACYCDuet™-1 is designed for the coexpression of two target genes. The vector encodes two multiple cloning sites (MCS) each of which is receded by a T7 promoter, lac operator and ribosome binding site (rbs). The vector also carries the P15A replicon, lacI gene and chloramphenicol resistance gene, this vector can be transformed into the same cell with pETDuet™-1, pCDFDuet™-1, and pRSFDuet™-1 or pCOLADuet™-1 in the appropriate host strain for the coexpression of up to eight target genes.

Specifications

Promoter T7
Content And Storage Store at or below -70°C.
Quantity 10 μg
For Use With (Application) Coexpression of two target genes

For Research Use Only