Learn More
MilliporeSigma™ Nutrient Media
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
Supplier:  MilliporeSigma™ MHA00P2SM
 View more versions of this product
Catalog No. MHA00P2SM
Only null left
Explore available promotions
Description
Description
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
  • Each dry powder lot for media preparation assayed for purity and compared to satisfactory lot
  • Each m-Endo broth for total coliform lot and m-FC broth for fecal coliform analysis lot certified in accordance with the Safe Drinking Water Act
  • Lot QC certificates included in each package as added assurance for critical procedures
Order Info
Nutrient media comes in liquid form, packaged in 2mL plastic ampules. Refrigerate as detailed on label.
Specifications
Specifications
Green
Yeast and Mold
United States
Testing drinks for yeast, mold
50 x 2 mL
2°C to 10°C
Liquid
Yeast/Mold Selective
Ampule
Product Suggestions
Product Suggestions