missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Nutrient Media
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
Supplier: MilliporeSigma™ MHA00BSM2
Description
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
- Each dry powder lot for media preparation assayed for purity and compared to satisfactory lot
- Each m-Endo broth for total coliform lot and m-FC broth for fecal coliform analysis lot certified in accordance with the Safe Drinking Water Act
- Lot QC certificates included in each package as added assurance for critical procedures
Order Info
Nutrient media comes in liquid form, packaged in 2mL plastic ampules. Refrigerate as detailed on label.
Specifications
| Orange/Yellow | |
| Brettanomyces species | |
| United States | |
| Brettanomyces testing | |
| 50 x 2 mL |
| 2°C to 10°C | |
| Liquid | |
| Brettanomyces Selective Broth | |
| Ampule |